
Heptaminol hydrochloride

Synonym(s):
6-Amino-2-methyl-2-heptanol hydrochloride, Heptaminol hydrochloride
Linear Formula:
CH3CH(NH2)(CH2)3C(OH)(CH3)2 · HCl
CAS Number:
Molecular Weight:
181.70
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing