Skip to Content
Merck
Heneicosanoic acid analytical standard

Heneicosanoic acid

Heneicosanoic acid analytical standard
Linear Formula:
CH3(CH2)19COOH
CAS Number:
Molecular Weight:
326.56
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing