Skip to Content
Merck
Guaiacol Impurity E Pharmaceutical Secondary Standard; Certified Reference Material

Guaiacol Impurity E

Guaiacol Impurity E Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Methyl benzoate
Linear Formula:
C6H5COOCH3
CAS Number:
Molecular Weight:
136.15

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1