Fulvestrant
Synonym(s):
(7α,17β)-7-[9-[(4,4,5,5,5-Pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-triene-3,17-diol, Faslodex, ICI 182,780, ZD 182780, ZD 9238, ZM 182780
Empirical Formula (Hill Notation):
C32H47F5O3S
CAS Number:
Molecular Weight:
606.77
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3