
Fmoc-Lys(Z)-OH

Synonym(s):
Nα-Fmoc-Nε-Z-L-lysine, Fmoc-Lys(Z)-OH
Empirical Formula (Hill Notation):
C29H30N2O6
CAS Number:
Molecular Weight:
502.56
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing