Skip to Content
Merck
Flunarizine dihydrochloride European Pharmacopoeia (EP) Reference Standard

Flunarizine dihydrochloride

Flunarizine dihydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
1-[Bis(4-fluorophenyl)methyl]-4-(3-phenyl-2-propenyl)piperazine dihydrochloride
Empirical Formula (Hill Notation):
C26H26F2N2 · 2HCl
CAS Number:
Molecular Weight:
477.42

Products

Product Category
Brand
Formula Weight
Manufacturer
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2