
Fenpropimorph

Empirical Formula (Hill Notation):
C20H33NO
CAS Number:
Molecular Weight:
303.48
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2