
Fenofibrate impurity A

Synonym(s):
4-Chloro-4′-hydroxybenzophenone, (4-Chlorophenyl)(4-hydroxyphenyl)methanone
Linear Formula:
ClC6H4COC6H4OH
CAS Number:
Molecular Weight:
232.66
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing