
Dulbecco’s Modified Eagle’s Medium/High Modified

Synonym(s):
Dulbecco’s Modified Eagle’s Medium - high glucose, DME, DMEM
Products
Product Category
Brand
Physical Form
Glucose (g/L)
L-Glutamine
Phenol Red
Sodium Pyruvate (g/L)
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing