
Diflufenican

Empirical Formula (Hill Notation):
C19H11F5N2O2
CAS Number:
Molecular Weight:
394.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2