
Diethofencarb

Synonym(s):
Isopropyl N-(3,4-diethoxyphenyl)carbamate
Empirical Formula (Hill Notation):
C14H21NO4
CAS Number:
Molecular Weight:
267.32
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2