
Diclofenac for system suitability

Synonym(s):
Diclofenac sodium salt, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt
Empirical Formula (Hill Notation):
C14H10Cl2NNaO2
CAS Number:
Molecular Weight:
318.13
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing