
Clotrimazole Impurity F

Synonym(s):
1-(Triphenylmethyl)imidazole
Empirical Formula (Hill Notation):
C22H18N2
CAS Number:
Molecular Weight:
310.39
Products
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing