Clenbuterol hydrochloride
Synonym(s):
4-Amino-α-(t-butylaminomethyl)-3,5-dichlorobenzyl alcohol hydrochloride
Empirical Formula (Hill Notation):
C12H18Cl2N2O · HCl
CAS Number:
Molecular Weight:
313.65
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3