
Chlorpropamide

Synonym(s):
1-(p-Chlorobenzenesulfonyl)-3-propylurea
Empirical Formula (Hill Notation):
C10H13ClN2O3S
CAS Number:
Molecular Weight:
276.74
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3