
Cefoperazone dihydrate

Empirical Formula (Hill Notation):
C25H27N9O8S2 · 2H2O
CAS Number:
Molecular Weight:
681.70
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3