
Buspirone Related Compound A

Synonym(s):
1-(2-Pyrimidyl)piperazine, 2-(1-Piperazinyl)pyrimidine
Empirical Formula (Hill Notation):
C8H12N4
Molecular Weight:
164.21
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing