
BSA for protein quantitation

Synonym(s):
Bovine Serum Albumin, Albumin bovine serum, BSA, Bovine albumin
CAS Number:
Products
Product Category
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing