
Bosentan

Synonym(s):
Bosentan monohydrate, 4-tert-Butyl-N-[6-(2-hydroxyethoxy)- 5-(2-methoxyphenoxy)-2-(pyrimidin-2-yl)pyrimidin-4-yl]benzenesulfonamide
Empirical Formula (Hill Notation):
C27H29N5O6S · H2O
CAS Number:
Molecular Weight:
569.63
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing