
β-Glucuronidase from limpets (Patella vulgata)

Synonym(s):
β-D-Glucuronide glucuronosohydrolase
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Recombinant Host
Quality Segment
Showing 1-3 of 3
Compare
Product Number
Description
Pricing