
β-Glucuronidase from Escherichia coli

Synonym(s):
β-D-Glucuronide glucuronosohydrolase
CAS Number:
Products
Brand
Biological Source
Physical Form
Recombinant Host
Chemical Composition
Quality Segment
Showing 1-6 of 6
Compare
Product Number
Description
Pricing