
Benzylpenicillin potassium

Synonym(s):
Penicillin G potassium salt, Benzylpenicillin potassium salt
Empirical Formula (Hill Notation):
C16H17KN2O4S
CAS Number:
Molecular Weight:
372.48
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing