
(−)-α-Thujone

Synonym(s):
(1S,4R)-1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one
Empirical Formula (Hill Notation):
C10H16O
Molecular Weight:
152.23
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing