
Anti-MLF2 antibody produced in rabbit

Synonym(s):
Anti-Myelodysplasia-myeloid leukemia factor 2 antibody produced in rabbit, Anti-Myeloid leukemia factor 2 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing