![Anti-FLAG® M2 Magnetic Beads affinity isolated antibody](/deepweb/assets/sigmaaldrich/product/images/235/147/235a2685-e5f0-44f0-b89f-9e6abf50f1c5/640/235a2685-e5f0-44f0-b89f-9e6abf50f1c5.jpg)
Anti-FLAG® M2 Magnetic Beads
![Anti-FLAG® M2 Magnetic Beads affinity isolated antibody](/deepweb/assets/sigmaaldrich/product/images/235/147/235a2685-e5f0-44f0-b89f-9e6abf50f1c5/640/235a2685-e5f0-44f0-b89f-9e6abf50f1c5.jpg)
Synonym(s):
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, Anti-ddddk, Anti-dykddddk
Products
Product Category
Brand
Clonality
Isotype
Physical Form
Analyte Suitability
Antibody Form
Conjugate
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing