
5-Amino-1,3-dihydro-2H-benzimidazol-2-one

Empirical Formula (Hill Notation):
C7H7N3O
CAS Number:
Molecular Weight:
149.15
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing