
5′-(4-Fluorosulfonylbenzoyl)adenosine hydrochloride

Synonym(s):
Adenosine-5′-(4-fluorosulfonylbenzoate) hydrochloride, FSBA
Empirical Formula (Hill Notation):
C17H16FN5O7S · HCl
CAS Number:
Molecular Weight:
489.86
Products
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing