![4-Vinylbenzyl chloride technical, ≥90% (GC), contains 0.1% total stabilizer](/deepweb/assets/sigmaaldrich/product/structures/213/610/85b7772c-2b74-44af-99c5-d16596f65c2f/640/85b7772c-2b74-44af-99c5-d16596f65c2f.png)
4-Vinylbenzyl chloride
![4-Vinylbenzyl chloride technical, ≥90% (GC), contains 0.1% total stabilizer](/deepweb/assets/sigmaaldrich/product/structures/213/610/85b7772c-2b74-44af-99c5-d16596f65c2f/640/85b7772c-2b74-44af-99c5-d16596f65c2f.png)
Synonym(s):
4-(Chloromethyl)styrene
Linear Formula:
H2C=CHC6H4CH2Cl
CAS Number:
Molecular Weight:
152.62
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing