![2-Nitrobenzaldehyde 98%](/deepweb/assets/sigmaaldrich/product/structures/370/821/4f86cbec-cac4-4401-a3c6-ae4ce95105f0/640/4f86cbec-cac4-4401-a3c6-ae4ce95105f0.png)
2-Nitrobenzaldehyde
![2-Nitrobenzaldehyde 98%](/deepweb/assets/sigmaaldrich/product/structures/370/821/4f86cbec-cac4-4401-a3c6-ae4ce95105f0/640/4f86cbec-cac4-4401-a3c6-ae4ce95105f0.png)
Synonym(s):
2-Nitrobenzaldehyde
Linear Formula:
2-(NO2)C6H4CHO
CAS Number:
Molecular Weight:
151.12
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing