
2-Ethyl-6-methoxynaphthalene

Synonym(s):
Ethylnerolin
Empirical Formula (Hill Notation):
C13H14O
CAS Number:
Molecular Weight:
186.25
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing