
2-Chloro-N,N-dimethylacetamide

Synonym(s):
N,N-Dimethylchloroacetamide, 2-Chloro-N,N-dimethylacetamide
Linear Formula:
ClCH2CON(CH3)2
CAS Number:
Molecular Weight:
121.57
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing