Skip to Content
Merck
2-(Boc-oxyimino)-2-phenylacetonitrile 99%

2-(Boc-oxyimino)-2-phenylacetonitrile

2-(Boc-oxyimino)-2-phenylacetonitrile 99%
Synonym(s):
2-(tert-Butoxycarbonyloxyimino)-2-phenyl­acetonitrile, Boc-ON
Linear Formula:
(CH3)3COCOON=C(C6H5)CN
CAS Number:
Molecular Weight:
246.26
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing