Mammalian Biologics Development and Manufacturing Services
OPTIMIZE YOUR BIOLOGIC DRUG DEVELOPMENT JOURNEY
With Millipore® CTDMO Services, we support biotech and biopharma companies developing a range of mammalian biologics including monoclonal antibodies (mAbs), mAbs for antibody-drug conjugates (ADCs), and mAb derived molecules such as bispecifics, antibody fragments and fc-fusion proteins.
Our CMC experts, dedicated development and MSAT teams provide specific core competencies to optimize process performance, drive greater cost efficiency, and ensure flexible mid-scale manufacturing across clinical stages and commercialization.
Our Track Record
35+
Years experience in process development
30
Years experience in GMP manufacturing
10+
Years experience in Single-use technology
280+
Projects (mAb, bsAb, fab, fc-fusion)
100+
Successful tech transfers
Development
Mammalian Process Development Services
Our dedicated development experts take a risk-based approach for an optimal balance between speed and cost, while maintaining highest quality standards. We support process development or process transfer and optimization of existing bioprocesses.
- Upstream and downstream process development
- Proven tech transfer of existing bioprocesses to our Mammalian Biologics Center of Excellence
- Process fitting and optimization
- Media and feed screening
- Streamlined downstream purification platform for increased performance and decreased process duration
- Drug substance formulation development
- Pilot lab capabilities up to 200L
- Direct process scale-up model from 3L to 2,000L
Mammalian Analytical Development Services
Our analytical development and validation teams support the entire product lifecycle with strong, phase-appropriate analytical capabilities.
- In-process control
- Platform methods for mAbs
- Multi-Attribute Method (MAM) platform
- Product-specific methods
- High-throughput methods
- Binding assays (ELISA, Biocore)
- Extended characterization
- HCP coverage & characterization
- Comprehensive mass spectrometry expertise
- Troubleshooting
TECHNOLOGY TRANSFER
Manufacturing Risk Assessment
Our CMC experts, dedicated development and MSAT teams work with our clients to identify gaps and mitigate risks for process fitting and evaluate potential process improvements to help reduce clinical and commercial manufacturing cost via process efficiencies. Through our manufacturing risk assessment, we:
- Assess critical quality attributes (CQA) and potential critical process parameters (pCPP) via FMEA
- Introduce platform processes and next-generation analytical methods for streamlined, scalable production
- Recommend adjustments to process design and required analytical methods validation
- Mitigate risks related to supply and compatibility of raw materials with commercial manufacturing
- Deliver a summary report with action plan and recommended CMC and tech transfer strategy for early and late-phase projects
Proven Technology Transfer
Throughout the transfer process, we effectively balance the factors for success including:
- CQA, CPP and process ranges (NOR, PAR)
- Process comparability and consistency
- Process reproducibility
- Process and analytical panel robustness
- CQA monitoring via our Multi-Attribute Method (MAM) platform for a smoother tech transfer and regulatory pathway
- Robust quality management system and ALCOA principales for documentation
- Flawless project management, detailed Gantt charts and open, transparent communication
MANUFACTURING
Mammalian GMP Manufacturing
Our flexible facility and capacity for clinical and commercial-scale production supports global drug substance supply for monoclonal antibodies (mAb), mAbs for Antibody-Drug Conjugates (ADCs), and mAb-derived molecules such as bispecifics, antibody fragments and fusion proteins.
- Mobius® single-use bioreactor platform
- Mid-scale manufacturing at 2,000 L
- Small-scale manufacturing at 200 L
- Strong Manufacturing, Science and Technology (MSAT) expertise
- Proven tech transfer
Commercial Manufacturing Readiness
Our process validation and MSAT teams have in-depth knowledge and scientific expertise of all aspects of upstream and downstream processing and validate the bioprocess for commercial-scale manufacturing.
- Risk assessment (FMEA) to define critical process parameters and process ranges (NOR, PAR)
- Scale-down model to gain process efficiencies before commercialization
- Pre-characterization before Process Design helps gain more data without impacting time to PPQ campaign
- Process characterization
- Robust control strategy (process, material, testing)
- Process Performance Qualification (PPQ)
- Resin lifetime study, resin reuse, mixing study and hold time study
- Continuous process verification
- BLA filing support
- Troubleshooting
REGULATORY SUPPORT
Tech Transfer Support
Our highly experienced regulatory experts are integral to each project, staying with it from the initial stages of the technology transfer through to scientific advice, regulatory filing and lifecycle management. We review and evaluate:
- Gap analysis & Chemistry, Manufacturing, and Controls (CMC) strategy
- Process control strategy
- Process changes post original
- Process and analytical methods fit-for-purpose
- GMP compliance
- Raw material origin
- Change notifications and regulatory impact
Scientific Advice
Review or author:
- Briefing books supporting meetings with Health Authorities (HA)
- Attendance of HA meetings upon request
- Responses to HA post submission
Regulatory Filing Support & Lifecycle Management
Review or author:
- CMC sections of regulatory dossier submissions(IND/IMPD & MMA/BLA)
- Information to the Common Technical Document (CTD) to build the CMC for the drug substances that will be submitted
- Support for post-approval changes
- Deviations and annual updates
- ROW continued filing support
MAMMALIAN BIOLOGIC PLATFORMS AND PACKAGES
Downstream Purification Platform
Biotech and biopharmaceutical companies can benefit from our streamlined platform to replace multiple polishing steps with a single flow-through step and decrease process duration and manufacturing cost. We optionally introduce our downstream purification platform for potential process improvements of existing bioprocesses.
We use mixed-mode chromatography to replace the typical two-step polishing of cation and anion exchange chromatography. Anionic mixed-mode chromatography combines anion exchange and hydrophobic interaction to remove all impurities including host cell proteins, DNA, and aggregates in a more efficient manner with equivalent quality.
Mammalian Analytical Packages
Our analytical packages support the entire product lifecycle, covering method development , transfer, optimization and analytical method validation. Our analytical experts consider our client’s program as a whole and ensure that each method is fit for purpose and compliant with local pharmacopeia and regulatory requirements. Several options are available for the development of the analytical panel:
An analytical package consisting of up-to-date, off-the-shelf techniques that assess content, purity, activity, and safety controls and includes platform methods such as ELISA, H/UPLC, qPCR, capillary electrophoresis, and glycan profile analysis can be leveraged. This templated, phase-appropriate strategy for monoclonal antibodies eliminates the time and expense of method development from scratch, while still allowing for flexibility.
For product-specific needs, methods can be custom-built by our analytical development team and transferred to our validation and QC labs. Our analytical capabilities includes methods such as binding assays (ELISA, SPR), enzymatic assays, SPR affinity & Kinetic (SPR), RP-U/HPLC, IEX-UPLC and others.
If a client has an existing set of analytical methods, they can be transferred as is, or optimized as part of the technology transfer process. This process may include adding or replacing current methods with state-of-the-art techniques to improve performance or throughput. Our high-throughput methods and equipment include OD HT microfluidic equipment, CE-SDS, cIEF, N-Glycan mapping using microfluidic, reverse CE-SDS by capillary, along with a pipetting robot for sample preparation.
Our analytical development team has extensive expertise in mass-spectrometry which clients can leverage for HCP characterization & quantification, peptide mapping, disulfide mapping, intact mass, and Multi-Attribute Method (MAM) platform to monitor post translational modifications and Critical Quality Attributes (CQA) in routine testing.
Multi-Attribute Method (MAM) Platform
Our MAM platform, based on liquid chromatography coupled to mass spectrometry (LC–MS), can be deployed to support biopharmaceutical companies at any stage, across process development, tech transfer and process validation. It’s a platform-based method that enables quality-by-design (QbD) and monitors product quality attributes (PQA) simultaneously, in one single assay.
It provides a consistent link between the process and the molecule PQAs, particularly important in process characterization studies, which aim at understanding the link between Critical Process Parameters (CPPs) and Critical Quality Attributes (CQAs) to refine process ranges for commercial manufacturing.
Fine-tuned through deep scientific expertise and years of experience, our MAM platform offers the high specificity, precision, accuracy, and robustness needed to make it the perfect tool to reliably monitor Critical Quality Attributes (CQA) and comply to regulatory requirements for smoother BLA submissions.
MAMMALIAN BIOLOGICS CENTER OF EXCELLENCE
Embedded into a global life science organization, we deliver Mammalian Biologics CDMO services across clinical phases and commercialization.
Established in 1987, we have supported monoclonal antibody (mAb) and mAb derivative development and production projects, since the industry’s beginning.
Our 2,700m2 manufacturing facility is equipped with 100% single-use technology for rapid scale up and flexible multi-product capacity.
Discover our journey to commercial manufacturing
When you work in this industry, you have a purpose, which is improving patients’ lives. With adding commercial manufacturing capabilties, this is exactly what we are doing.
Flexible facility design
In this video interview you will learn how we have leveraged latest industry practices to build a flexible facility.
Related Product Resources
- Why Choosing a Client-centric CTDMO is Critical when Outsourcing
This infographic discoveres 5 considerations, that are most critical as indicators of success.
- Trends in Outsourcing the Development and Manufacturing of Biologics
In this whitepaper, we share highlights from a global market research survey of small, midsized and large biopharmaceutical companies on trends related to outsourcing.
- A Molecule’s Journey Break Down Roadblocks to Clinical Success
A guidebook for today’s biopharma executives seeking to navigate through the important considerations necessary to successfully bring a molecule to the clinic.
- Optimize the Formulation for a Monoclonal Antibody
This case study shows, how our innovative, custom solutions and decades of expertise allow to effectively balance cost, speed and risk when optimizing drug substance formulations in an accelerated timeframe.
- Reduce the Level of Host Cell Proteins in the Bioreactor Harvest
Learn how conceptualized and effective approaches help to address process development challenges such as HCP removal, in an accelerated timeframe.
- Scale your Process Directly from 3L to 2000L
In this case study we showcase how to advance a drug candidate into the clinic and ultimately onto the market faster.
- A Molecule’s Journey: Break Down Roadblocks to Commercial Manufacturing Success
A guidebook for today’s biopharma executives outlining key considerations to help ensure commercial manufacturing success.