Alcohols are typically compatible with this product. Strong acids and bases are not recommended. See the image below to review the product compatibility table:
Z742648
Tube-O-DIALYZER™ mini dialysis system
Micro, MWCO 1 K
Synonym(s):
semi-permeable membrane tube, single use tube dialysis
Select a Size
CLP 75,010
Estimated to ship onApril 26, 2025
Select a Size
About This Item
CLP 75,010
Estimated to ship onApril 26, 2025
Recommended Products
feature
Micro
packaging
pkg of 5 ea
manufacturer/tradename
G-BioSciences 786-610T
pore size
1 K MWCO
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
The Tube-O-DIALYZER tube comes in two ideal sizes; the Micro unit allows efficient dialysis of 20-250μl samples and the Medi unit is optimized for samples between 0.2-2.5ml. Both sizes are available with membranes with molecular weight cutoffs (MWCO) of 1k, 4k, 8k, 15k, and 50k Daltons.
Tube-O-DIALYZER comes in packs of 20, each supplied with 6 appropriately sized floats and storage caps to allow storage of dialyzed samples. For added convenience, Tube-O-DIALYZER is also available as a mixed kit containing 10 Micro and 10 Medi units.
Application
- Dialysis of small sample volumes
- Equilibrium dialysis for buffer exchange
- Dialysis for single use applications, such as infectious and radioactive samples
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Will organic solution like ethanol cause potential damage to this dialysis membrane?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service