Z617822
Greiner tissue culture tube with screw cap
capacity 14 mL
All Photos(1)
About This Item
Recommended Products
material
round bottom transparent polystyrene
screw top cap
sterility
sterile
packaging
pack of 1,000 ea
manufacturer/tradename
Greiner 163160
capacity
14 mL
diam. × L
16 mm × 100 mm
General description
Greiner CELLSTAR® is manufactured from crystal clearpolystyrene and has improved cell adhesion through physical surface treatment.Non-skirted, supplied with a red screw cap.
Features and Benefits
- Sterile
- Free detectable DNase, human DNA, RNase
- Non cytotoxic and non pyrogenic
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH