Skip to Content
Merck
All Photos(1)

Key Documents

SAB5200102

Sigma-Aldrich

Anti-SLC9A3 antibody produced in rabbit

1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-NHE3, Anti-Slc9a3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 84 kDa

species reactivity

rat, mouse

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Solute carrier family 9 member A3 (SLC9A3) is also known as Na+/H+ exchanger 3 (NHE3) and is expressed in the brush border epithelium. The gene ID of the protein is 24784.

Specificity

Detects ~84 kDa.

Immunogen

AA809 to 831 of rat sequence

Biochem/physiol Actions

Solute carrier family 9 member A3 (SLC9A3) is an exchanger which takes part in the reabsorption of water and sodium chloride in the proximal tubule of kidneys. It exchanges one Na+ (from the lumen) with one H+ (from the cytosol).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jens Christian Brasen et al.
American journal of physiology. Renal physiology, 307(11), F1249-F1262 (2014-10-10)
The proximal tubule Na(+)/H(+) exchanger 3 (NHE3), located in the apical dense microvilli (brush border), plays a major role in the reabsorption of NaCl and water in the renal proximal tubule. In response to a rise in blood pressure NHE3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service