MAK550
Tyrosinase Assay Kit
sufficient for 100 colorimetric tests
About This Item
Recommended Products
application(s)
cosmetics
food and beverages
detection method
colorimetric
relevant disease(s)
cancer
shipped in
wet ice
storage temp.
−20°C
General description
The Tyrosinase Assay Kit is a simple, one-step and reliable assay for monitoring tyrosinase activity with high sensitivity. The assay uses a colorless substrate solution that significantly increases its absorption at 510 nm upon reaction with tyrosinase. The increases in absorption at 510 nm is proportionate to the tyrosinase activity.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service