Skip to Content
Merck
All Photos(1)

Key Documents

CLS4868

Corning® universal fit pipet tips

100-1000 μL universal fit bulk packed pipet tips, blue, non-sterile, 1000 tips/bag, 1000 tips/case

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
volume range:
100-1000 μL
sterility:
non-sterile
feature:
DNA free: no
RNase free: no
barrier: no
disposable
endotoxin free
filter: no
Bulk Pack
packaging:
bag of 1000
case of 1000
manufacturer/tradename:
Corning 4868

material

blue tip
polypropylene

sterility

non-sterile

feature

DNA free: no
RNase free: no
barrier: no
disposable
endotoxin free
filter: no
Bulk Pack

packaging

bag of 1000
case of 1000

manufacturer/tradename

Corning 4868

volume range

100-1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

Corning® Universal Fit Pipet Tips

Corning Universal Fit ploypropylene pipet tips are designed to provide a reliable fit with all major brand pipettors and have a bevelled orifice to help ensure fluid delivery. All racked tips are certified RNase/DNase free and nonpyrogenic.

  • Reliable fit with all major brand pipettors
  • Bevelled orifice to help ensure fluid delivery
  • Certified RNase/DNase free and nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service