Skip to Content
Merck
All Photos(1)

Key Documents

MOPC 315

85022106, mouse blood, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MOPC 315, 85022106

biological source

mouse blood

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

Immunoglobulin A lambda

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse myeloma

Cell Line Description

Established in vitro from the MOPC 315 plasmacytoma induced by mineral oil injections in a BALB/c mouse. Secretes an IgA lambda antibody which recognises DNP-TNP substituted proteins. Cells are killed by low concentrations (10nM) of dexamethasone.

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10-20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 1-9x100,000 cells/ml; 5% CO2; 37°C. Do not allow cell density to rise above 1,000,000 cells/ml as rapid death occurs at higher concentrations.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service