Skip to Content
Merck
All Photos(1)

Key Documents

46300

Supelco

Ethoxysulfuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N4O7S
CAS Number:
Molecular Weight:
398.39
Beilstein:
9284797
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

CCOc1ccccc1OS(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2

InChI

1S/C15H18N4O7S/c1-4-25-10-7-5-6-8-11(10)26-27(21,22)19-15(20)18-14-16-12(23-2)9-13(17-14)24-3/h5-9H,4H2,1-3H3,(H2,16,17,18,19,20)

InChI key

UWVKRNOCDUPIDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Qi et al.
Mikrochimica acta, 187(5), 279-279 (2020-04-22)
A "half" core-shell g-C3N4/Fe3O4@ZIF-8 nanohybrid, in which Fe3O4 and zeolite imidazolate framework-8 (ZIF-8) constructed the core-shell structure, was successfully fabricated via a versatile in situ growth strategy. This nanohybrid was employed for simultaneous magnetic solid-phase extraction (MSPE) of trace levels
Fábio Gozzi et al.
Journal of the science of food and agriculture, 96(5), 1580-1584 (2015-05-15)
Due to contamination of the environment by pesticides and their mishandling, there is the need for treatment of contaminated sites and correct disposal of materials containing them. Thus, studies with advanced oxidation processes are expanding and can determine the rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service