24365C
EX-CELL® CD CHO Fusion
Animal-component free, chemically defined cell culture medium, without L-glutamine, without hypoxanthine, without sodium bicarbonate, without thymidine
Pharma Manufacturing
About This Item
Recommended Products
Quality Level
product line
EX-CELL®
form
dry powder
technique(s)
cell culture | mammalian: suitable (suspension)
shipped in
ambient
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Quantity
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Cellvento 4CHO-X Expansion Medium enhances cell biomass for bioreactor inoculation in cell expansion stages.
Cellvento® and EX-CELL® CHO cell culture media and feeds, optimized for monoclonal antibody & recombinant protein production in fed-batch & perfusion
Related Content
Learn more about cell line development and the relevant products you need for this process.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service