Skip to Content
Merck
All Photos(1)

Key Documents

CDRC050HG

Milli-Q

Reverse Osmosis Cartridge

For Reverse Osmosis at a flowrate of 50 L/h, For use with Elix® and RiOs water purification systems

Synonym(s):

Reverse Osmosis Cartridge – 50 L/h

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104214
NACRES:
JA.13

packaging

pkg of 1 unit

compatibility

for use with Elix®
for use with RiOs

Looking for similar products? Visit Product Comparison Guide

General description

The reverse osmosis (RO) membrane removes 95%-99% of contaminants including ions, particulates, bacteria, and organic molecules. For use with Elix® 35/70/100 and RiOs 50/100/150/200 water purification systems.

Application

The RO water delivered from RiOs and Elix® systems is used in laboratories, mainly for applications such as general rinsing, feedwater to washing machines and autoclaves, or as a feed to pure and ultrapure water purification systems.

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service