8.18784
3-Nonanol
for synthesis
Synonym(s):
3-Nonanol, Ethyl hexyl carbinol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Quality Level
Assay
≥96.0% (GC)
bp
192-194 °C
transition temp
flash point 96 °C
density
0.82 g/cm3 at 20 °C
storage temp.
2-30°C
InChI
1S/C9H20O/c1-3-5-6-7-8-9(10)4-2/h9-10H,3-8H2,1-2H3
InChI key
GYSCXPVAKHVAAY-UHFFFAOYSA-N
Application
3-Nonanol for synthesis. CAS No. 624-51-1, EC Number 210-850-6.
Analysis Note
Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.823 - 0.826
Identity (IR): passes test
Density (d 20 °C/ 4 °C): 0.823 - 0.826
Identity (IR): passes test
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
204.8 °F - closed cup
Flash Point(C)
96 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service