Skip to Content
Merck
All Photos(1)

Key Documents

8.04015

Sigma-Aldrich

Furfuryl alcohol

for synthesis

Synonym(s):

Furfuryl alcohol, 2-Furylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-626-1
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.53 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

390 °C

potency

177 mg/kg LD50, oral (Rat)
400 mg/kg LD50, skin (Rabbit)

expl. lim.

1.8-16.3 % (v/v)

pH

6 (20 °C, 300 g/L in H2O)

bp

170-171 °C/1013 mbar

mp

-31 °C

transition temp

flash point 65 °C

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[o]1c(ccc1)CO

InChI

1S/C5H6O2/c6-4-5-2-1-3-7-5/h1-3,6H,4H2

InChI key

XPFVYQJUAUNWIW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.131 - 1.133
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Nose, Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service