Skip to Content
Merck
All Photos(1)

Key Documents

8.03467

Sigma-Aldrich

Dichloromethyl methyl ether

for synthesis

Synonym(s):

Dichloromethyl methyl ether, 1,1-Dichloromethylether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4Cl2O
CAS Number:
Molecular Weight:
114.96
MDL number:
UNSPSC Code:
12352112
EC Index Number:
225-498-9
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

84 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

bp

85-87 °C/1013 hPa

transition temp

flash point 42 °C

density

1.29 g/cm3 at 20 °C

SMILES string

ClC(Cl)OC

InChI

1S/C2H4Cl2O/c1-5-2(3)4/h2H,1H3

InChI key

GRTGGSXWHGKRSB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service