Skip to Content
Merck
All Photos(1)

Key Documents

8.01776

Sigma-Aldrich

Benzoin

for synthesis

Synonym(s):

Benzoin, α-Hydroxy-α-phenylacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)COC6H5
CAS Number:
Molecular Weight:
212.24
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-331-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.3 hPa ( 136 °C)

Quality Level

Assay

≥99.0% (GC)

form

crystals

potency

10000 mg/kg LD50, oral (Rat)
8870 mg/kg LD50, skin (Rabbit)

bp

344 °C/1023 hPa

mp

132-134 °C

transition temp

flash point 181 °C

solubility

0.3 g/L

density

1.31 g/cm3 at 20 °C

bulk density

350 kg/m3

storage temp.

no temp limit

SMILES string

OC(c2ccccc2)C(=O)c1ccccc1

InChI

1S/C14H12O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13,15H

InChI key

ISAOCJYIOMOJEB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 130 °C
Melting range (upper value): ≤ 136 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service