Skip to Content
Merck
All Photos(1)

Documents

GF04691460

Calcium

lump, 10 mm max. lump size, weight 50 g, purity 99%

Synonym(s):

CA006101

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141503
NACRES:
NA.23

Assay

99%

form

lumps

manufacturer/tradename

Goodfellow 046-914-60

weight

50 g

max. lump size

10 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service