Skip to Content
Merck
All Photos(3)

Key Documents

523682

Sigma-Aldrich

Chitosan oligosaccharide lactate

average Mn 5,000

Synonym(s):

Polyvinylsulfuric acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H24N2O9)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

mol wt

average Mn 5,000

Quality Level

composition

oligosaccharide, 60%

SMILES string

N[C@H]1[C@H](O)O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2N)[C@@H]1O

InChI

1S/C12H24N2O9/c13-5-9(19)10(4(2-16)21-11(5)20)23-12-6(14)8(18)7(17)3(1-15)22-12/h3-12,15-20H,1-2,13-14H2/t3-,4-,5-,6-,7-,8-,9-,10-,11-,12+/m1/s1

InChI key

QLTSDROPCWIKKY-PMCTYKHCSA-N

General description

Mn = 4,000 - 6,000

Application

Chitosan oligosaccharide lactate may be used in water treatment for organic and heavy metal pollutants, biomedical devices, biodegradable packaging, textile finishing for water permeable fabrics, cosmetics.

Physical form

> 90% deacetylated

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nenad L Ignjatović et al.
Journal of materials chemistry. B, 6(43), 6957-6968 (2018-01-01)
Low targeting efficiency and fast metabolism of antineoplastic drugs are hindrances to effective chemotherapies and there is an ongoing search for better drugs, but also better carriers. Steroid derivatives, 3β-hydroxy-16-hydroxymino-androst-5-en-17-one (A) and 3β,17β-dihydroxy-16-hydroxymino-androst-5-ene (B) as cancer growth inhibitors were chemically
Yuly A Henao-Díaz et al.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc, 30(5), 671-677 (2018-07-22)
Routine collection and testing of oral fluid (OF) samples facilitates porcine reproductive and respiratory syndrome virus (PRRSV) surveillance in commercial swine herds in a cost-effective, welfare-friendly fashion. However, OFs often contain environmental contaminants that may affect liquid handling and test
Steinunn Gudmundsdottir et al.
Journal of biomedical materials research. Part A, 103(8), 2778-2785 (2015-02-17)
Chitosan, the deacetylated derivative of chitin, is used as biomaterial in diverse settings. It is also found on pathogens and can be proinflammatory. Shorter derivatives of chitosan can be generated chemically or enzymatically, chitosan oligosaccharides (ChOS). There is variation in

Articles

Polymers, natural or synthetic, are vital in medical advancements, contributing to device and pharmaceutical development in therapy.

Chitin, a natural polysaccharide, is the second most abundant natural biopolymer in the world, after cellulose.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service