Skip to Content
Merck
All Photos(1)

Key Documents

393231

Sigma-Aldrich

Dichloro[rac-ethylenebis(indenyl)]zirconium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16Cl2Zr
CAS Number:
Molecular Weight:
418.47
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: zirconium
reagent type: catalyst

impurities

≤3% dichloromethane

SMILES string

Cl[Zr]Cl.C(CC1C=Cc2ccccc12)C3C=Cc4ccccc34

InChI

1S/C20H18.2ClH.Zr/c1-3-7-19-15(5-1)9-11-17(19)13-14-18-12-10-16-6-2-4-8-20(16)18;;;/h1-12,17-18H,13-14H2;2*1H;/q;;;+2/p-2

InChI key

IJSDUKZKUCXMRL-UHFFFAOYSA-L

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organometallics, 10, 1501-1501 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service