Skip to Content
Merck
All Photos(1)

Key Documents

270954

Sigma-Aldrich

Tris(2,2,2-trifluoroethyl) phosphite

99%

Synonym(s):

Tris(2,2,2-trifluoroethoxy)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CH2O)3P
CAS Number:
Molecular Weight:
328.07
Beilstein:
1714041
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.324 (lit.)

bp

130-131 °C/743 mmHg (lit.)

density

1.487 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)COP(OCC(F)(F)F)OCC(F)(F)F

InChI

1S/C6H6F9O3P/c7-4(8,9)1-16-19(17-2-5(10,11)12)18-3-6(13,14)15/h1-3H2

InChI key

CBIQXUBDNNXYJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(2,2,2-trifluoroethyl) phosphite is an organophosphorus compound that is used as a ligand in organometallic chemistry to enhance labialization in metal clusters and as a substitutional equivalent for carbon monoxide. It is also used as an electrolyte additive to improve the electrochemical performance and as a co-solvent for nonflammable electrolytes in Li-ion batteries.

Application

Tris(2,2,2-trifluoroethyl) phosphite was used in the synthesis of N-methoxy-N-methyl bis(2,2,2-trifluoroethyl)phosphonamide via the Arbuzov reaction with 2-bromo-N-methoxy-N-methylacetamide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Fortin et al.
The Journal of organic chemistry, 67(15), 5437-5439 (2002-07-20)
The N-methoxy-N-methyl bis(2,2,2-trifluoroethyl)phosphonamide was easily obtained via the Arbuzov reaction with use of commercially available tris(2,2,2-trifluoroethyl)phosphite, 2-bromo-N-methoxy-N-methylacetamide, and KF/alumina. The reaction of bis(2,2,2-trifluoroethyl)phosphonate with several aldehydes demonstrates the versatility of the method, which gives Z-unsaturated amides in good yields.
Jiulin Wang et al.
Angewandte Chemie (International ed. in English), 53(38), 10099-10104 (2014-07-26)
Of the various beyond-lithium-ion batteries, lithium-sulfur (Li-S) batteries were recently reported as possibly being the closest to market. However, its theoretically high energy density makes it potentially hazardous under conditions of abuse. Therefore, addressing the safety issues of Li-S cells
Krzysztof Matyjaszewski et al.
Molecules (Basel, Switzerland), 26(2) (2021-01-14)
N-alkyl phosphoranimines were synthesized via the Staudinger reaction of four different alkyl azides with tris(2,2,2-trifluoroethyl) phosphite. N-adamantyl, N-benzyl, N-t-butyl, and N-trityl phosphoranimines were thoroughly characterized and evaluated as chain-capping compounds in the anionic polymerization of P-tris(2,2,2-trifluoroethoxy)-N-trimethylsilyl phosphoranimine monomer. All four

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service